Tricin Puffersubstanz
Lieferant: G Bioscienes
Synonyme: N-(Tris(hydroxymethyl)methyl)glycin
RC-104 RC-103
RC-104EA 735 EUR
RC-104 RC-103
Tricin Puffersubstanz
Tricin
This tricine is a buffer component used for the separation of low molecular weight peptides.
Formel: (HOCH₂)₃CNHCH₂CO₂H
Molecular Weight: 179,17 g/mol
Schmelzpunkt: 187 °C
Dichte: 1,523 g/cm³ (19.85 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004277
CAS-Nummer: 5704-04-1
EINECS: 227-193-6
Merck Index: 13,09724