Gas washing bottles heads
Lieferant: Lenz Laborglas GmbH & CO.KG
05530301 05530300
LENZ05530301EA 49.2 EUR
LENZ05530301 LENZ05530300
Gas washing bottles heads
Verschlüsse Flaschenverschlüsse
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes