Wash bottles, narrow neck, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F116210008 F116210016
BELAF116210008EA 280 EUR
BELAF116210008 BELAF116210016
Wash bottles, narrow neck, SP Bel-Art
Flaschen Spritzflaschen
LDPE, with PP cap und delivery tube
  • Simple to squeeze
  • Extra fine needle spray
  • Good chemical resistance, good temperature resistance
Ultra thin spray for precise aiming.