Permanent marker pen, Edding 800
Lieferant: THEODOR KIHLER BURO EINRICHTUN
800-1 800-2 800-3 800-4
811-3101EA 25.2 EUR
811-3101 811-3102 811-3103 811-3104
Permanent marker pen, Edding 800
Schreibstifte Permanentmarker
Permananent marker for writing, colouring, labelling for materials such as metal, glass or plastic, low odour ink, round interchangeable tip. Line thickness of 4 to 12 mm.
  • Fast drying
  • Waterproof
  • Refillable