Anti-TRIOBP Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP34718_P050EA 385 EUR
AVIVARP34718_P050
Anti-TRIOBP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TRIOBP (TRIO and F-actin binding protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR