Anti-SLC12A2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP43805_T100EA 335 EUR
AVIVARP43805_T100
Anti-SLC12A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC12A2 (solute carrier family 12 (sodium/potassium/chloride transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR