Anti-THEG Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP57809_P050EA 385 EUR
AVIVARP57809_P050
Anti-THEG Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: THEG (theg spermatid protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR