Anti-CDC26 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP59256_P050EA 385 EUR
AVIVARP59256_P050
Anti-CDC26 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CDC26 (cell division cycle 26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR