Anti-SYT5 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP59484_P050EA 385 EUR
AVIVARP59484_P050
Anti-SYT5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SYT5 (synaptotagmin V)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR