Anti-C1QA Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP60064_P050EA 385 EUR
AVIVARP60064_P050
Anti-C1QA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: C1QA (complement component 1, q subcomponent, A chain)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR