Anti-RAB1B Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP64096_P050EA 385 EUR
AVIVARP64096_P050
Anti-RAB1B Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAB1B (RAB1B, member RAS oncogene family)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR