Anti-NUDT14 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVARP67709_P050EA 385 EUR
AVIVARP67709_P050
Anti-NUDT14 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NUDT14 (nudix (nucleoside diphosphate linked moiety X)-type motif 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR