Anti-GABRP Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
AVIVAVARP13034P050EA 385 EUR
AVIVAVARP13034P050
Anti-GABRP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GABRP (gamma-aminobutyric acid (GABA) A receptor, pi)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen