Anti-YBX3 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVIVP100916_T100EA 335 EUR
AVIVP100916_T100
Anti-YBX3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: YBX3 (Y box binding protein 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR