Anti-RPS6KB2 Mouse Monoclonal Antibody

Lieferant: Abgent
ABGEAM1854BEA 348 EUR
ABGEAM1854B
Anti-RPS6KB2 Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: RPS6KB2 (ribosomal protein S6 kinase, 70kDa, polypeptide 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR