Anti-NSUN3 Rabbit Polyclonal Antibody [clone: 164CT21.2.2]

Lieferant: Abgent
ABGEAP11287AEA 348 EUR
ABGEAP11287A
Anti-NSUN3 Rabbit Polyclonal Antibody [clone: 164CT21.2.2]
Antikörper
Type: Primary
Antigen: NSUN3 (NOP2/Sun domain family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR