Anti-PCCA Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP12328CEA 348 EUR
ABGEAP12328C
Anti-PCCA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PCCA (propionyl CoA carboxylase, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR