Anti-SLC27A3 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP13457AEA 348 EUR
ABGEAP13457A
Anti-SLC27A3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC27A3 (solute carrier family 27 (fatty acid transporter), member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR