Anti-PTK2 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP8614BEA 348 EUR
ABGEAP8614B
Anti-PTK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTK2 (protein tyrosine kinase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR