Anti-MARC1 Rabbit Polyclonal Antibody

Lieferant: Abgent
ABGEAP9754CEA 348 EUR
ABGEAP9754C
Anti-MARC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MARC1 (mitochondrial amidoxime reducing component 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR