Anti-EXOC7 Goat polyclonal antibody

Lieferant: OriGene
ORIGTA303105EA 547 EUR
ORIGTA303105
Anti-EXOC7 Goat polyclonal antibody
Antikörper
Recommended Dilutions: ELISA: 1:16000; Western Blot: 05-15µg/ml; Immunochemistry: 5-10µg/ml

Type: Primary
Antigen: EXOC7 (exocyst complex component 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR