Anti-AGR2 Goat Polyclonal Antibody

Lieferant: OriGene
ORIGTA303161EA 547 EUR
ORIGTA303161
Anti-AGR2 Goat Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:128000; Western Blot: up to 001µg/ml

Type: Primary
Antigen: AGR2 (anterior gradient 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR