Anti-RAMP2 Mouse Monoclonal Antibody [clone: OTI4E5]

Lieferant: OriGene
ORIGTA504410EA 598 EUR
ORIGTA504410
Anti-RAMP2 Mouse Monoclonal Antibody [clone: OTI4E5]
Antikörper
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: RAMP2 (receptor (G protein-coupled) activity modifying protein 2)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR