Anti-HDAC1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB1767-100EA 907 EUR
ABCAAB1767-100
Anti-HDAC1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: HDAC1 (histone deacetylase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR