Anti-Bovine Serum Albumin Mouse Monoclonal Antibody [clone: 5F9]
Lieferant: Abcam
ABCAAB9092-100EA 882 EUR
ABCAAB9092-100
Anti-Bovine Serum Albumin Mouse Monoclonal Antibody [clone: 5F9]
Antikörper
Type: Primary
Antigen: Bovine Serum Albumin
Clonality: monoclonal
Clone: 5F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Bovine