Anti-IgG1 Mouse Monoclonal Antibody [clone: MARG1-2] (Biotin)
Lieferant: Abcam
ABCAAB11668-250EA 396 EUR
ABCAAB11668-250
Anti-IgG1 Mouse Monoclonal Antibody [clone: MARG1-2] (Biotin)
Antikörper
Type: Secondary
Antigen: Rat IgG1
Clonality: monoclonal
Clone: MARG1-2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen