Anti-Histone H1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB17347-100EA 793 EUR
ABCAAB17347-100
Anti-Histone H1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Histone H1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR