Anti-Fibronectin Mouse monoclonal antibody unconjugated [clone: Fn-3]

Lieferant: Abcam
ABCAAB18265-50EA 902 EUR
ABCAAB18265-50
Anti-Fibronectin Mouse monoclonal antibody unconjugated [clone: Fn-3]
Antikörper
Type: Primary
Antigen: Fibronectin (fibronectin)
Clonality: monoclonal
Clone: Fn-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR