Anti-VPS26A Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB23892-100EA 1030 EUR
ABCAAB23892-100
Anti-VPS26A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VPS26A (vacuolar protein sorting 26 homolog A (S. pombe))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat