Anti-CYPIVF11 Mouse Monoclonal Antibody [clone: F21 P6 F5]

Lieferant: Abcam
ABCAAB55841-100EA 837 EUR
ABCAAB55841-100
Anti-CYPIVF11 Mouse Monoclonal Antibody [clone: F21 P6 F5]
Antikörper
Type: Primary
Antigen: CYPIVF11
Clonality: monoclonal
Clone: F21 P6 F5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR