Anti-PIKFYVE Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB62467-100EA 837 EUR
ABCAAB62467-100
Anti-PIKFYVE Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PIKFYVE (phosphoinositide kinase, FYVE finger containing)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse