Anti-XRCC5 Rabbit Monoclonal Antibody [clone: EPR3467]

Lieferant: Abcam
ABCAAB79391-100EA 933 EUR
ABCAAB79391-100
Anti-XRCC5 Rabbit Monoclonal Antibody [clone: EPR3467]
Antikörper
Type: Primary
Antigen: XRCC5 (X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining))
Clonality: monoclonal
Clone: EPR3467
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR