Anti-CRTC2 Rabbit monoclonal antibody unconjugated [clone: EPR3384]

Lieferant: Abcam
ABCAAB109081-100EA 900 EUR
ABCAAB109081-100
Anti-CRTC2 Rabbit monoclonal antibody unconjugated [clone: EPR3384]
Antikörper
Type: Primary
Antigen: CRTC2 (CREB regulated transcription coactivator 2)
Clonality: monoclonal
Clone: EPR3384
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR