Anti-ITPR1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB111615-100EA 882 EUR
ABCAAB111615-100
Anti-ITPR1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ITPR1 (inositol 1,4,5-trisphosphate receptor, type 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR