Anti-SIRT6 Mouse Monoclonal Antibody [clone: 1G3]

Lieferant: Abcam
ABCAAB119007-100EA 811 EUR
ABCAAB119007-100
Anti-SIRT6 Mouse Monoclonal Antibody [clone: 1G3]
Antikörper
Type: Primary
Antigen: SIRT6 (sirtuin 6)
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, African green monkey, Dog
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR