Anti-ACTA2 Mouse Monoclonal Antibody [clone: 4A4]

Lieferant: Abcam
ABCAAB119952-100EA 719 EUR
ABCAAB119952-100
Anti-ACTA2 Mouse Monoclonal Antibody [clone: 4A4]
Antikörper
Type: Primary
Antigen: ACTA2 (actin, alpha 2, smooth muscle, aorta)
Clonality: Monoclonal
Clone: 4A4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, African green monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR