Anti-BMP4 Rabbit Monoclonal Antibody [clone: EPR6211]

Lieferant: Abcam
ABCAAB124715-100EA 902 EUR
ABCAAB124715-100
Anti-BMP4 Rabbit Monoclonal Antibody [clone: EPR6211]
Antikörper
Type: Primary
Antigen: BMP4 (bone morphogenetic protein 4)
Clonality: monoclonal
Clone: EPR6211
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR