Anti-NDRG3 Rabbit Monoclonal Antibody [clone: EPR9010]

Lieferant: Abcam
ABCAAB131266-100EA 825 EUR
ABCAAB131266-100
Anti-NDRG3 Rabbit Monoclonal Antibody [clone: EPR9010]
Antikörper
Type: Primary
Antigen: NDRG3 (NDRG family member 3)
Clonality: monoclonal
Clone: EPR9010
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR