Anti-SERPINE2 Rabbit Monoclonal Antibody
Lieferant: Abcam
ABCAAB134905-100EA 922 EUR
ABCAAB134905-100
Anti-SERPINE2 Rabbit Monoclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINE2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen