Anti-IgG Goat Polyclonal Antibody (CF™ 488A)

Lieferant: Biotium

20012.EA 289 EUR
20012. 20012-1
Anti-IgG Goat Polyclonal Antibody (CF™ 488A)
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR