Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Lieferant: Biotium
20014-1EA 144 EUR
20014-1 20014.
Anti-IgG Donkey Polyclonal Antibody (CF™ 488A)
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 488A Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Mouse