Anti-IgG Goat Polyclonal Antibody (CF™ 647)

Lieferant: Biotium

20043.EA 289 EUR
20043. 20043-1
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 647 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR