Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody [clone: PyMT]

Lieferant: Novus Biologicals
NOVUNB100-2749EA 610 EUR
NOVUNB100-2749 NOVUNB100-2749SS
Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody [clone: PyMT]
Antikörper
Type: Primary
Antigen: Polyoma Virus, Medium T Antigen
Clonality: monoclonal
Clone: PyMT
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Virus
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR