Anti-CCR4 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB100-56336EA 466 EUR
NOVUNB100-56336
Anti-CCR4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CCR4 (chemokine (C-C motif) receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR