Anti-ATF2 Mouse monoclonal antibody unconjugated [clone: 103C411.2]

Lieferant: Novus Biologicals
NOVUNB100-56594EA 586 EUR
NOVUNB100-56594
Anti-ATF2 Mouse monoclonal antibody unconjugated [clone: 103C411.2]
Antikörper
Type: Primary
Antigen: ATF2 (activating transcription factor 2)
Clonality: monoclonal
Clone: 103C411.2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR