Anti-TOM70 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB110-58347EA 580 EUR
NOVUNB110-58347 NOVUNB110-58347SS
Anti-TOM70 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TOM70
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR