Anti-MMP2 Mouse Monoclonal Antibody (DyLight® 350) [clone: 2C1]

Lieferant: Novus Biologicals
NOVUNB200-113UVEA 696 EUR
NOVUNB200-113UV
Anti-MMP2 Mouse Monoclonal Antibody (DyLight® 350) [clone: 2C1]
Antikörper
Type: Primary
Antigen: MMP2 (matrix metallopeptidase 2 (gelatinase A, 72kDa gelatinase, 72kDa type IV collagenase))
Clonality: monoclonal
Clone: 2C1
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Guinea pig, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR