Anti-HIST3H3 Rabbit Polyclonal Antibody
Lieferant: Novus Biologicals
NOVUNB21-1081EA 601 EUR
NOVUNB21-1081 NOVUNB21-1081SS
Anti-HIST3H3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIST3H3 (histone cluster 3, H3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen