Anti-SMAD5 Rabbit Polyclonal Antibody

Lieferant: Novus Biologicals
NOVUNB600-962EA 546 EUR
NOVUNB600-962
Anti-SMAD5 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SMAD5 (SMAD family member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR