Anti-STRO-1 Mouse Monoclonal Antibody [clone: STRO-1]

Lieferant: Novus Biologicals
NOVUNBP1-48356SSEA 237 EUR
NOVUNBP1-48356SS NOVUNBP1-48356
Anti-STRO-1 Mouse Monoclonal Antibody [clone: STRO-1]
Antikörper
Type: Primary
Antigen: STRO-1
Clonality: monoclonal
Clone: STRO-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM Lambda
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR